Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF323 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589728
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589728 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589728 Supplier Invitrogen™ Supplier No. PA589728
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDSKLQRDVS LDSKYRETCK RDSKAEKQQA HSTGERRHRC NECGKSFTKS SVLIEHQRIH TGEKPYECEE CGKAFSRRSS LNEHRRSHTG EKPYQCKECG KAFSASNGLT RHRRIHTGEK PYECKVCGKA FLLSSCLVQH QRIHTGEKRY QCRECGKAFI QNAGLFQHLR VHTGEKPYQC SQCSKLFSKR TLLKKHQKIH TGERP; Positive Samples: Rat liver; Cellular Location: Nucleus.

ZNF323 is a member of the subfamily of C2H2 Kruppel-like zinc finger transcription factors that have a SCAN box domain (Pi et al., 2002 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF323
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Alias dJ874C20.2; zinc finger and SCAN domain containing 31; zinc finger and SCAN domain-containing protein 31; Zinc finger protein 323; ZNF20-Lp; ZNF310P; ZNF323; ZSCAN31
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 202-406 of human ZSCAN31 (NP_1121613).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.