Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF324 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558925
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558925 Supplier Invitrogen™ Supplier No. PA558925
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HTGEKTVRRS RASLHPQARS VAGASSEGAP AKETEPTPAS GPAAVSQPAE Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 38%.

ZNF324 belongs to the krueppel C2H2-type zinc-finger protein family.It contains 9 C2H2-type zinc fingers and 1 KRAB domain. ZNF324 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF324
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF324
Gene Accession No. O75467
Gene Alias ZF5128; zinc finger protein 324; zinc finger protein 324A; Zinc finger protein ZF5128; ZNF324; ZNF324A
Gene Symbols ZNF324
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF324. Recombinant protein control fragment (Product #RP-98349).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25799
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.