Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF324B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554791
Description
Immunogen sequence: KPFVCTQCGR AFRERPALLH HQRIHTTEKT NAAAPDCTPG PGFLQGHHRK VRRGGKPSPV LKPAKV Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 43%.
May be involved in transcriptional regulation.
Specifications
ZNF324B | |
Polyclonal | |
Unconjugated | |
ZNF324B | |
Zinc finger protein 324B; ZNF324B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
388569 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6AW86 | |
ZNF324B | |
Recombinant protein corresponding to Human ZNF324B. Recombinant protein control fragment (Product #RP-93604). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction