Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF326 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541698
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541698 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541698 Supplier Invitrogen™ Supplier No. PA541698
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GNIQGVGEGG EVGVVGEVEG VGEVEEVEEL EEETAKEEPA DFPVEQPEEN Sequence homology: Dog: 79%; Human: 100%; Rat: 93%.

Core component of the DBIRD complex, a multiprotein complex that acts at the interface between core mRNP particles and RNA polymerase II (RNAPII) and integrates transcript elongation with the regulation of alternative splicing: the DBIRD complex affects local transcript elongation rates and alternative splicing of a large set of exons embedded in (A + T)-rich DNA regions. May play a role in neuronal differentiation and is able to bind DNA and activate expression in vitro. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF326
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF326
Gene Accession No. Q5BKZ1
Gene Alias 5730470H14Rik; DBIRD complex subunit ZNF326; dJ871E2.1; dJ871E2.1 (novel protein (ortholog of mouse zinc finger protein Zan75)); Zan75; Zfp326; zinc finger protein 326; zinc finger protein 326; DBIRD complex subunit ZNF326; zinc finger protein interacting with mRNPs; Zinc finger protein interacting with mRNPs and DBC1; zinc finger protein-associated with nuclear matrix of 75 kDa; Zird; ZNF326; ZNF-protein interacting with nuclear mRNPs and DBC1
Gene Symbols ZNF326
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ZNF326 (aa 533-582).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284695
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.