Learn More
ZNF326 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | ZNF326 |
---|---|
Applications | Immunofluorescence, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1982
![]() |
Bethyl Laboratories, Inc
IHC00371T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1981
![]() |
Bethyl Laboratories, Inc
IHC00371 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Monkey, Gorilla, Chimpanzee, Northern white-cheeked gibbon and Crab-eating macaque
Probable transcriptional activator which may play a role in neuronal differentiation. Able to bind DNA and activate expression in vitro.Specifications
ZNF326 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
ZAN75, Zfp326, dJ871E2.1, dJ871E2.1 (novel protein (ortholog of mouse zinc finger protein Zan75)) | |
ZNF326 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q5BKZ1 | |
284695 | |
Between 532 and 582 | |
Primary | |
4° C | |
ZNF326 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.