Learn More
ZNF326 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301880A
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
Probable transcriptional activator which may play a role in neuronal differentiation. Able to bind DNA and activate expression in vitro.Specifications
| ZNF326 | |
| Polyclonal | |
| Unconjugated | |
| ZNF326 | |
| ZAN75, Zfp326, dJ871E2.1, dJ871E2.1 (novel protein (ortholog of mouse zinc finger protein Zan75)) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 284695 | |
| 4° C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q5BKZ1 | |
| ZNF326 | |
| Between 532 and 582 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.