Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF329 Monoclonal Antibody (OTI2F3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA809544
Description
May be involved in transcriptional regulation.Specifications
| ZNF329 | |
| Monoclonal | |
| Unconjugated | |
| ZNF329 | |
| zinc finger protein 329 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79673 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI2F3 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q86UD4 | |
| ZNF329 | |
| Human recombit protein fragment corresponding to amino acids 1-203 of human ZNF329 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction