Learn More
ZNF329 Mouse anti-Human, Clone: OTI2D12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809582

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
May be involved in transcriptional regulation.Specifications
ZNF329 | |
Monoclonal | |
Unconjugated | |
ZNF329 | |
zinc finger protein 329 | |
Mouse | |
Affinity Chromatography | |
RUO | |
79673 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2D12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q86UD4 | |
ZNF329 | |
Human recombit protein fragment corresponding to amino acids 1-203 of human ZNF329 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.