Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF329 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531903
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531903 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531903 Supplier Invitrogen™ Supplier No. PA531903
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, MCF-7. Predicted reactivity: Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Zinc finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc finger proteins contain a Kruppeltype DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. As a member of the Krueppel C2H2-type zinc finger protein family, ZNF329 (zinc finger protein 329) is a 541 amino acid nuclear protein that contains twelve C2H2-type zinc fingers through which it is thought to be involved in DNA-binding and transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF329
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ZNF329
Gene Accession No. Q86UD4
Gene Alias 2810439M05Rik; 4632409L22Rik; RGD1565029; Zfp329; Zinc finger protein 329; Znf329
Gene Symbols ZNF329
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZNF329. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79673
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.