Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF329 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560693
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560693 Supplier Invitrogen™ Supplier No. PA560693
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EHLIASSDLP PSQRVLATNG FHAPDSNVSG LDCDPALPSY PKSYADKRTG DSDACGKGFN HSMEVIHGR Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 48%.

Zinc finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc finger proteins contain a Kruppeltype DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. As a member of the Krueppel C2H2-type zinc finger protein family, ZNF329 (zinc finger protein 329) is a 541 amino acid nuclear protein that contains twelve C2H2-type zinc fingers through which it is thought to be involved in DNA-binding and transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF329
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF329
Gene Accession No. Q86UD4
Gene Alias 2810439M05Rik; 4632409L22Rik; RGD1565029; Zfp329; Zinc finger protein 329; Znf329
Gene Symbols ZNF329
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF329. Recombinant protein control fragment (Product #RP-93629).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79673
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.