Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF330 Monoclonal Antibody (OTI8E9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA810145

Description
The function of this protein remains unknown.Specifications
ZNF330 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9Y3S2 | |
ZNF330 | |
Full length human recombit protein of human ZNF330 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI8E9 | |
Unconjugated | |
ZNF330 | |
HSA6591, NOA36 | |
Mouse | |
Affinity Chromatography | |
RUO | |
27309 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction