Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF330 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553367
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553367 Supplier Invitrogen™ Supplier No. PA553367
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SCQVLEAETF KCVSCNRLGQ HSCLRCKACF CDDHTRSKVF KQEKGKQPPC PKCGHETQET KDLSMSTRSL KFGRQTGGEE GDGASGYDAY WKNLSSDKYG DTSYHD Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF330
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF330
Gene Accession No. Q9Y3S2
Gene Alias BC008086; HSA6591; NOA 36; Noa36; Nucleolar autoantigen 36; nucleolar cysteine-rich protein; Zfp330; zinc finger 330-like protein; zinc finger autoantigen 330; zinc finger protein 330; zinc finger protein 330-like protein; Znf330
Gene Symbols ZNF330
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NOA36. Recombinant protein control fragment (Product #RP-90726).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27309
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.