Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF331 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566560
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566560 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566560 Supplier Invitrogen™ Supplier No. PA566560
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NIHEIRASKRN SDRRSKSLGR NWICEGTLER PQRSRGRYVN QMIINYVKRP ATREGTPPRT HQRHHKENSF Highest antigen sequence identity to the following orthologs - mouse 31%, rat 31%.

ZNF331 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 12 C2H2-type zinc fingers and 1 KRAB domain. ZNF331 may be involved in transcriptional regulation. It may play a role in spermatogenesis. Zinc finger proteins have been shown to interact with nucleic acids and to have diverse functions. The zinc finger domain is a conserved amino acid sequence motif containing 2 specifically positioned cysteines and 2 histidines that are involved in coordinating zinc. Kruppel-related proteins form one family of zinc finger proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF331
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF331
Gene Accession No. Q9NQX6
Gene Alias C2H2-like zinc finger protein rearranged in thyroid adenomas; DKFZp686L0787; KRAB zinc finger protein; RITA; Zinc finger protein 331; zinc finger protein 361; Zinc finger protein 463; ZNF331; ZNF361; ZNF463
Gene Symbols ZNF331
Host Species Rabbit
Immunogen Recombinant Human ZNF331. Recombinant protein control fragment (Product #RP-105256).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55422
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.