Learn More
Invitrogen™ ZNF331 Polyclonal Antibody

Description
ZNF331 Polyclonal Antibody for ICC/IF, IHC (P), ELISA
Specifications
Specifications
| Antigen | ZNF331 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | ZNF331 |
| Gene Accession No. | Q9NQX6 |
| Gene Alias | C2H2-like zinc finger protein rearranged in thyroid adenomas; DKFZp686L0787; KRAB zinc finger protein; RITA; Zinc finger protein 331; zinc finger protein 361; Zinc finger protein 463; ZNF331; ZNF361; ZNF463 |
| Gene Symbols | ZNF331 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.