Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF333 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145169
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145169 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145169 Supplier Invitrogen™ Supplier No. PA5145169
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QCQPQEAIPS QDTFTEILSI DVKGEQPQPG EKLYKYNELE KPFNSIEPLF QYQRIHAGEA SCECQEIRNS FFQSAHLIVP EKIRSGDKSY ACNKCE Antigen sequence identity: Mouse 36%; Rat 36%.

ZNF333 belongs to the krueppel C2H2-type zinc-finger protein family. Itcontains 10 C2H2-type zinc fingers and 2 KRAB domains. ZNF333 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF333
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF333
Gene Accession No. Q96JL9
Gene Alias KIAA1806; Zinc finger protein 333; ZNF333
Gene Symbols ZNF333
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 277-372. Recombinant protein control fragment (Product #RP-109940).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84449
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.