Learn More
Invitrogen™ ZNF333 Polyclonal Antibody

Description
Immunogen sequence: MESVTFEDVA VEFIQEWALL DSARRSLCKY RMLDQCRTLA SRGTPPCKPS CVSQLGQRAE PKATERGILR ATGVAWESQL KPEELPSMQD LLEEASSRDM QMGPGLFLRM QLVPSIEERE TPLTREDRPA LQEPPWSLGC TGLKAAMQIQ RVVIPVPTLG HRNPWVARDS AVPARDPAWL QEDKVEEEAM APGLPTACSQ EPVTFADVAV VFTPEEWVFL DSTQRSLYRD VMLENYRNLA SVADQLCKPN ALSYLEERGE QWTTDRGVLS DTCAEPQCQP QEAIPSQDTF TEILSIDVKG PPN; Positive Samples: HeLa, Mouse ovary, Rat brain, Rat heart; Cellular Location: Nucleus.
Specifications
Specifications
| Antigen | ZNF333 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.45 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | ZNF333 |
| Gene Accession No. | Q96JL9 |
| Gene Alias | KIAA1806; Zinc finger protein 333; ZNF333 |
| Gene Symbols | ZNF333 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.