Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF335 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566653
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566653 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566653 Supplier Invitrogen™ Supplier No. PA566653
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RNGHLKFHIQR LHSPDGRKSG TPTARAPTQT PTQTIILNSD DETLATLHTA LQSSHGVLGP ERLQQALSQE HIIVAQEQ Highest antigen sequence identity to the following orthologs - mouse 85%, rat 82%.

Component or associated component of some histone methyltransferase complexes may regulate transcription through recruitment of those complexes on gene promoters. Enhances ligand-dependent transcriptional activation by nuclear hormone receptors. Plays an important role in neural progenitor cell proliferation and self-renewal through the regulation of specific genes involved brain development, including REST. Also controls the expression of genes involved in somatic development and regulates, for instance, lymphoblast proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF335
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF335
Gene Accession No. Q9H4Z2
Gene Alias 1810045J01Rik; LOW QUALITY PROTEIN: zinc finger protein 335; MCPH10; Nif; NIF1; NIF-1; NIF2; NRC-interacting factor 1; Zfp335; Zinc finger protein 335; zinc-finger/leucine-zipper co-transducer NIF1; zinc-finger/leucine-zipper co-transducer NIF-1; Znf335
Gene Symbols ZNF335
Host Species Rabbit
Immunogen Recombinant Human ZNF335. Recombinant protein control fragment (Product #RP-107304).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63925
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.