Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF335 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598258
Description
ZNF335 Polyclonal Antibody for Western Blot, ELISA
Component or associated component of some histone methyltransferase complexes may regulate transcription through recruitment of those complexes on gene promoters. Enhances ligand-dependent transcriptional activation by nuclear hormone receptors. Plays an important role in neural progenitor cell proliferation and self-renewal through the regulation of specific genes involved brain development, including REST. Also controls the expression of genes involved in somatic development and regulates, for instance, lymphoblast proliferation.
Specifications
| ZNF335 | |
| Polyclonal | |
| Unconjugated | |
| ZNF335 | |
| 1810045J01Rik; LOW QUALITY PROTEIN: zinc finger protein 335; MCPH10; Nif; NIF1; NIF-1; NIF2; NRC-interacting factor 1; Zfp335; Zinc finger protein 335; zinc-finger/leucine-zipper co-transducer NIF1; zinc-finger/leucine-zipper co-transducer NIF-1; Znf335 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 63925 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 6.025 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9H4Z2 | |
| ZNF335 | |
| Recombinant Human Zinc finger protein 335 protein (672-909AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction