Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF337 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565380
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565380 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565380 Supplier Invitrogen™ Supplier No. PA565380
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YSHLVSLGIYA EHVLRPKNLG LAHQRQQQLQ FSDQSFQSDT AEGQEKEKST KPMAFSSPPL RHAVSSRRRN SVVEIESSQG QRENPTEIDK VLKGIENSRW GA Highest antigen sequence identity to the following orthologs - mouse 23%, rat 27%.

This gene encodes a zinc finger domain containing protein. The function of this protein has yet to be determined. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Mar 2014].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF337
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF337
Gene Accession No. Q9Y3M9
Gene Alias Zinc finger protein 337; ZNF337
Gene Symbols ZNF337
Host Species Rabbit
Immunogen Recombinant Human ZNF337. Recombinant protein control fragment (Product #RP-104791).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26152
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.