Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF33A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563335
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563335 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563335 Supplier Invitrogen™ Supplier No. PA563335
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VWTADHLKER SQENQSKHLW EVVFINNEML TKEQ Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 34%.

ZNF33A belongs to the krueppel C2H2-type zinc-finger protein family. It contains 16 C2H2-type zinc fingers and 1 KRAB domain. ZNF33A may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF33A
Applications ChIP Assay, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF33A
Gene Accession No. Q06730
Gene Alias brain my041 protein; KIAA0065; KOX2; KOX31; KOX5; NF11A; Zinc finger and ZAK-associated protein with KRAB domain; zinc finger protein 11A; Zinc finger protein 33A; zinc finger protein 33a (KOX 31); zinc finger protein KOX31; ZNF11; ZNF11A; ZNF33; ZNF33A; ZZAPK
Gene Symbols ZNF33A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF33A. Recombinant protein control fragment (Product #RP-104509).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7581
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.