Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF34 Antibody (OTI3G2), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274937F
Description
ZNF34 Monoclonal specifically detects ZNF34 in Human samples. It is validated for Immunohistochemistry.Specifications
ZNF34 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
ZNF34 | |
Recombinant protein fragment corresponding to amino acids 117-405 of human ZNF34 (NP_085057) produced in E.coli. | |
0.1 mL | |
80778.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI3G2 | |
Immunohistochemistry | |
FLJ3472, KOX21zinc finger protein 37a (KOX 21), zinc finger protein 37A, Zinc finger protein KOX21, ZNF37 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction