Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF34 Monoclonal Antibody (OTI3G2)
GREENER_CHOICE

Catalog No. PIMA526608
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526608 Supplier Invitrogen™ Supplier No. MA526608
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF34 Monoclonal Antibody for Western Blot, IHC (P)

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF34
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3G2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZNF34
Gene Accession No. Q8IZ26
Gene Alias KOX32; Zinc finger protein 34; zinc finger protein 34 (KOX 32); zinc finger protein KOX32; ZNF34
Gene Symbols ZNF34
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 117-405 of ZNF34 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80778
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.