Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF34 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563892
Description
Immunogen sequence: QETFGEEDPQ GSEPVEACDH ISKSEGSLEK LVEQRGPRAV TLTNGESSRE SGGNLRLL Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 31%.
May be involved in transcriptional regulation.
Specifications
ZNF34 | |
Polyclonal | |
Unconjugated | |
ZNF34 | |
KOX32; Zinc finger protein 34; zinc finger protein 34 (KOX 32); zinc finger protein KOX32; ZNF34 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80778 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IZ26 | |
ZNF34 | |
Recombinant protein corresponding to Human ZNF34. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction