Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF34 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566868
Description
Immunogen sequence: LSRPVPDQRPH KCDICEQSFE QRSYLNNHTR VHRS Highest antigen sequence identity to the following orthologs - mouse 46%, rat 46%.
May be involved in transcriptional regulation.
Specifications
| ZNF34 | |
| Polyclonal | |
| Unconjugated | |
| ZNF34 | |
| KOX32; Zinc finger protein 34; zinc finger protein 34 (KOX 32); zinc finger protein KOX32; ZNF34 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80778 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IZ26 | |
| ZNF34 | |
| Recombinant Human ZNF34. Recombinant protein control fragment (Product #RP-107519). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction