Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF341 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584745
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584745 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584745 Supplier Invitrogen™ Supplier No. PA584745
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVWPPGHSGG TVSRNSVTVQ VMALNPSRQE DEESTGLGQP LPGAPQPQAL STAGEEEGDK PESKQVVLID SSY Highest antigen sequence indentity to the following orthologs: Rat - 30%, Mouse - 84%.

The ZNF341 gene is located on chromosome 20.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF341
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF341
Gene Accession No. Q9BYN7
Gene Alias dJ553F4.3; ZFP341; zinc finger protein 341; ZNF341
Gene Symbols ZNF341
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF341. Recombinant protein control fragment (Product #RP-105416).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84905
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.