Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF341 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540309
Description
Peptide sequence: GEEEGDKPES KQVVLIDSSY LCQFCPSKFS TYFQLKSHMT QHKNEQVYKC Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Mouse: 93%; Rabbit: 86%.
The ZNF341 gene is located on chromosome 20.
Specifications
| ZNF341 | |
| Polyclonal | |
| Unconjugated | |
| ZNF341 | |
| dJ553F4.3; ZFP341; zinc finger protein 341; ZNF341 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84905 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BYN7 | |
| ZNF341 | |
| Synthetic peptide directed towards the middle region of human ZNF341. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction