Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF342 Antibody

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP288676
Description
ZNF342 Polyclonal specifically detects ZNF342 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ZNF342 | |
Polyclonal | |
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
MRX89MGC8941, zinc finger protein 41 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of human ZNF342 | |
100 μL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
162979 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction