Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF346 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232230AF405
Description
ZNF346 Polyclonal antibody specifically detects ZNF346 in Human samples. It is validated for Western BlotSpecifications
ZNF346 | |
Polyclonal | |
Western Blot | |
KIAA0844ZNF365D, MGC41821, MGC87345, Su48, talanin, UANprotein ZNF365, zinc finger protein 365 | |
The immunogen this antibody was made to, maps to a region between residue 244 to 294 of human Just Another Zinc Finger Protein using the numbering given in entry NP_036411.1 (GeneID 23567). | |
0.1 mL | |
Apoptosis | |
23567 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction