Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF346 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982061
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2061 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2061 Supplier Bioss Supplier No. BS12235R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Probable transcriptional activator which may play a role in neuronal differentiation. Able to bind DNA and activate expression in vitro.

Specifications

Antigen ZNF346
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZNF346
Gene Accession No. Q9UL40
Gene Alias double-stranded RNA-binding zinc finger protein JAZ; JAZ; Just another zinc finger protein; Zfp346; Zinc finger protein 346; ZNF346
Gene Symbols ZNF346
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZNF346.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23567
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.