Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF35 Antibody (OTI4D5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274938H
Description
ZNF35 Monoclonal specifically detects ZNF35 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ZNF35 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
ZNF35 | |
Recombinant protein fragment corresponding to amino acids 1-258 of human ZNF35 (NP_003411) produced in E.coli. | |
0.1 mL | |
7584.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4D5 | |
Western Blot, Immunohistochemistry | |
BRF2, Butyrate response factor 2, ERF2ERF-2, Protein TIS11D, RNF162Czinc finger protein 36, C3H1 type-like 2, TIS11DEGF-response factor 2, zinc finger protein 36, C3H type-like 1, zinc finger protein 36, C3H type-like 2, zinc finger protein, C3H type, 36-like 2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction