Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF35 Monoclonal Antibody (OTI3H5)
GREENER_CHOICE

Catalog No. PIMA526844
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526844 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526844 Supplier Invitrogen™ Supplier No. MA526844
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF35 Monoclonal Antibody for Western Blot, IHC (P)

ZNF35 is a new candidate transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF35
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3H5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZNF35
Gene Accession No. P13682
Gene Alias HF.10; HF10; mszf13; mszf20-1; mszf28; mszf68; Zfp105; Zfp239; Zfp35; Zfp-35; zinc finger protein 239; zinc finger protein 271; Zinc finger protein 35; zinc finger protein 35 (clone HF.10); zinc finger protein HF.10; Znf271; ZNF35
Gene Symbols ZNF35
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-258 of ZNF35 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7584
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.