Learn More
Invitrogen™ ZNF350 Polyclonal Antibody

Description
A suggested positive control is A-20 cell lysate. PA5-20614 can be used with blocking peptide PEP-0734.
Specifications
Specifications
| Antigen | ZNF350 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ZNF350 |
| Gene Accession No. | Q9GZX5 |
| Gene Alias | KRAB zinc finger protein ZFQR; ZBRK1; ZFQR; Zinc finger and BRCA1-interacting protein with a KRAB domain 1; zinc finger protein 350; Zinc finger protein ZBRK1; zinc-finger protein ZBRK1; ZNF350 |
| Gene Symbols | ZNF350 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.