Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF354A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562540
Description
Immunogen sequence: TQDSSFQGLI LKRSNRNVPW DLKLEKPYIY EGRLEKKQDK KGSFQIVSAT HKKIPTIERS HKNTELSQNF SPKSVLIRQQ ILPR Highest antigen sequence identity to the following orthologs: Mouse - 40%, Rat - 40%.
ZNF354A gene ontology annotations related to this gene include regulation of transcription by RNA polymerase II; sensory perception of sound.
Specifications
ZNF354A | |
Polyclonal | |
Unconjugated | |
ZNF354A | |
AW488485; epididymis luminal protein 104; EZNF; HEL104; HKL1; Kid1; KID-1; Kidney 1; kidney, ischemia, and developmentally-regulated protein 1; Renal transcription factor Kid-1; Tcf17; TCF-17; Transcription factor 17; Zfp354a; Zinc finger protein 354A; zinc finger protein 354B; zinc finger protein eZNF; ZNF354A; ZNF354B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6940 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60765 | |
ZNF354A | |
Recombinant protein corresponding to Human ZNF354A. Recombinant protein control fragment (Product #RP-101457). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction