Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF354A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568594
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568594 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568594 Supplier Invitrogen™ Supplier No. PA568594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 77%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.

ZNF354A gene ontology annotations related to this gene include regulation of transcription by RNA polymerase II; sensory perception of sound.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF354A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF354A
Gene Accession No. O60765
Gene Alias AW488485; epididymis luminal protein 104; EZNF; HEL104; HKL1; Kid1; KID-1; Kidney 1; kidney, ischemia, and developmentally-regulated protein 1; Renal transcription factor Kid-1; Tcf17; TCF-17; Transcription factor 17; Zfp354a; Zinc finger protein 354A; zinc finger protein 354B; zinc finger protein eZNF; ZNF354A; ZNF354B
Gene Symbols ZNF354A
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ZNF354A.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6940
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.