Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF354A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501729191 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9191 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9191 Supplier Proteintech Group Inc Supplier No. 140721AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF354A gene ontology annotations related to this gene include regulation of transcription by RNA polymerase II; sensory perception of sound.

Specifications

Antigen ZNF354A
Applications Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZNF354A
Gene Accession No. O60765, Q02975, Q61751
Gene Alias EZNF, HKL1, KID 1, KID1, TCF 17, TCF17, Transcription factor 17, zinc finger protein 354A, Zinc finger protein eZNF, ZNF354A
Gene Symbols ZNF354A
Host Species Rabbit
Immunogen ZNF354A Fusion Protein Ag5142
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21408, 24522, 6940
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.