Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF358 Antibody (PCRP-ZNF358-1A6), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314233R
This item is not returnable.
View return policy
Description
ZNF358 Monoclonal antibody specifically detects ZNF358 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
ZNF358 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
140467 | |
Store at 4°C in the dark. | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence | |
PCRP-ZNF358-1A6 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
hKID3, KID3Kidney, ischemia, and developmentally-regulated protein 3, KRAB-zinc finger protein synten, zinc finger protein 354C | |
Recombinant full-length human ZNF358protein (Uniprot: Q9NW07) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction