Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF358 Antibody (PCRP-ZNF358-1A6), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314233F
This item is not returnable.
View return policy
Description
ZNF358 Monoclonal antibody specifically detects ZNF358 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
ZNF358 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
140467 | |
Store at 4°C in the dark. | |
Purified |
Flow Cytometry, Immunofluorescence | |
PCRP-ZNF358-1A6 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
hKID3, KID3Kidney, ischemia, and developmentally-regulated protein 3, KRAB-zinc finger protein synten, zinc finger protein 354C | |
Recombinant full-length human ZNF358protein (Uniprot: Q9NW07) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction