Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF358 Mouse anti-Human, Clone: PCRP-ZNF358-1A6, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314233100UG
This item is not returnable.
View return policy
Description
ZNF358 Monoclonal antibody specifically detects ZNF358 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Protein ArraySpecifications
ZNF358 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug per 100-500 ug protein, Protein Array | |
hKID3, KID3Kidney, ischemia, and developmentally-regulated protein 3, KRAB-zinc finger protein synten, zinc finger protein 354C | |
Recombinant full-length human ZNF358protein (Uniprot: Q9NW07) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2c |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Peptide Array | |
PCRP-ZNF358-1A6 | |
Unconjugated | |
10mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
140467 | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction