Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF364 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10041373
Description
ZNF364 Polyclonal specifically detects ZNF364 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
ZNF364 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
E3 ubiquitin-protein ligase RNF115, EC 6.3.2.-, Rabring 7, ring finger protein 115CL469780, Zinc finger protein 364BCA2, ZNF364 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog, Pig and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:32000 | |
NP_055270.1 | |
RNF115 | |
Peptide with sequence C-DQDNRANERGHQTH corresponding to internal region according to NP_055270.1. | |
0.1 mg | |
Breast Cancer, Cancer, Zinc Finger | |
27246 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction