Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF115 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542304
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542304 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542304 Supplier Invitrogen™ Supplier No. PA542304
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PWLELHDTCP VCRKSLNGED STRQSQSTEA SASNRFSNDS QLHDRWTF Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 86%; Rabbit: 93%; Rat: 100%.

ZNF364 contains 1 RING-type zinc finger. The exact function of ZNF364 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF115
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RNF115
Gene Accession No. Q9Y4L5
Gene Alias 2610028E05Rik; AU042696; BCA2; CL469780; E3 ubiquitin-protein ligase RNF115; Rab7-interacting RING finger protein; rabring 7; Rabring7; ring finger protein 115; RING-type E3 ubiquitin transferase RNF115; RNF115; Zfp364; Zinc finger protein 364; ZNF364
Gene Symbols RNF115
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ZNF364 (aa 257-304).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27246
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.