Learn More
Invitrogen™ ZNF365 Monoclonal Antibody (2E3)

Description
Peptide Sequence: GLPTSDTKAS FEAHVREKFN RMVEAVDRTI EKRIDKLTKE LAQKTAELLE VRAAFVQLTQ KKQEVQRRER ALN.
Specifications
Specifications
| Antigen | ZNF365 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2E3 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZNF365 |
| Gene Accession No. | Q70YC5 |
| Gene Alias | AI839779; AV340892; DBZ; DISC1-binding zinc-finger protein; KIAA0844; LOW QUALITY PROTEIN: protein ZNF365; protein su48; Protein ZNF365; protein ZNF365-like; Su48; Talanin; UAN; Zfp365; zinc finger protein 365; ZNF365; ZNF365D |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.