Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF365 Monoclonal Antibody (OTI8G7), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA505049

Encompass_Preferred

 View more versions of this product

Catalog No. 50-167-5604


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

This gene encodes several isoforms which have different expression patterns and functions. Mutation in this gene is associated with uric acid nephrolithiasis (UAN). Alternatively spliced variants, encoding distinct proteins, have been identified.
Specifications

Specifications

ZNF365
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q70YC4
ZNF365
Human recombit protein fragment corresponding to amino acids 108-407 of human ZNF365 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Western Blot
OTI8G7
Unconjugated
ZNF365
Su48, UAN, ZNF365D
Mouse
Affinity Chromatography
RUO
22891
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.