Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF365 Monoclonal Antibody (OTI8G7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA505049S


Description
ZNF365 Monoclonal antibody specifically detects ZNF365 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
ZNF365 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q15714 | |
Mouse | |
Affinity chromatography | |
RUO | |
8848 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI8G7 | |
Unconjugated | |
Tsc22d1 | |
AI839779; AV340892; DBZ; DISC1-binding zinc-finger protein; KIAA0844; LOW QUALITY PROTEIN: protein ZNF365; protein su48; Protein ZNF365; protein ZNF365-like; Su48; Talanin; UAN; Zfp365; zinc finger protein 365; ZNF365; ZNF365D | |
Full length human recombinant protein of human TSC22D1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction