Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF365 Monoclonal Antibody (OTI9H5), TrueMAB™, OriGene
SDP

Catalog No. 502539940
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9940 30 μL
50-167-5615 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9940 Supplier OriGene Supplier No. TA505065S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZNF365 Monoclonal antibody specifically detects ZNF365 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ZNF365
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI9H5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Tsc22d1
Gene Accession No. Q15714
Gene Alias AI839779; AV340892; DBZ; DISC1-binding zinc-finger protein; KIAA0844; LOW QUALITY PROTEIN: protein ZNF365; protein su48; Protein ZNF365; protein ZNF365-like; Su48; Talanin; UAN; Zfp365; zinc finger protein 365; ZNF365; ZNF365D
Host Species Mouse
Immunogen Full length human recombinant protein of human TSC22D1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8848
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.