Learn More
ZNF365 Mouse anti-Human, Clone: OTI3B1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800111

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes several isoforms which have different expression patterns and functions. Mutation in this gene is associated with uric acid nephrolithiasis (UAN). Alternatively spliced variants, encoding distinct proteins, have been identified.Specifications
ZNF365 | |
Monoclonal | |
Unconjugated | |
ZNF365 | |
Su48, UAN, ZNF365D | |
Mouse | |
Affinity Chromatography | |
RUO | |
22891 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3B1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q70YC4 | |
ZNF365 | |
Human recombit protein fragment corresponding to amino acids 108-407 of human ZNF365 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.