Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Talanin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116312
Description
Talanin Polyclonal Antibody for Western Blot
Talanin may play a role in uric acid excretion.
Specifications
| Talanin | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q70YC4 | |
| LOC124902436 | |
| KLH conjugated synthetic peptide between 50-77 amino acids from the Central region of human ZNF365. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| LOC124902436 | |
| DBZ; DISC1-binding zinc-finger protein; KIAA0844; Protein su48; Protein ZNF365; Su48; Talanin; UAN; zinc finger protein 365; ZNF365; ZNF365D | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 124902436 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction