Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF367 Polyclonal Antibody

ZNF367 Polyclonal Antibody
Supplier: Thermo Scientific PA5107320
Description
ZNF367 Polyclonal Antibody for Western Blot
Specifications
| ZNF367 | |
| Polyclonal | |
| Unconjugated | |
| ZNF367 | |
| 8030486J21Rik; AFF29; C2H2 zinc finger protein ZFF29; C2H2 zinc finger protein ZFF29a; CDC14 cell division cycle 14 homolog B; CDC14B; habp4; hyaluronan binding protein 4; hypothetical protein LOC550421; LOW QUALITY PROTEIN: zinc finger protein 367; ZFF29; ZFF29a; ZFF29b; Zfp367; zgc:112157; Zinc finger protein 367; Znf367 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 195828 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q7RTV3 | |
| ZNF367 | |
| Synthetic peptide directed towards the C-terminal region of human ZNF367. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction