Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF367 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569497
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569497 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569497 Supplier Invitrogen™ Supplier No. PA569497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 85%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.

Transcriptional activator. Isoform 1 may be involved in transcriptional activation of erythroid genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF367
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Zfp367
Gene Accession No. Q5U2Z0
Gene Alias 8030486J21Rik; AFF29; C2H2 zinc finger protein ZFF29; C2H2 zinc finger protein ZFF29a; CDC14 cell division cycle 14 homolog B; CDC14B; habp4; hyaluronan binding protein 4; hypothetical protein LOC550421; LOW QUALITY PROTEIN: zinc finger protein 367; ZFF29; ZFF29a; ZFF29b; Zfp367; zgc:112157; Zinc finger protein 367; Znf367
Gene Symbols Zfp367
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of rat ZNF367.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306695
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.