Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF379 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987120
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7120 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7120 Supplier Bioss Supplier No. BS6200R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The ZDHHC9-GOLGA7 complex is a palmitoyltransferase specific for HRAS and NRAS.

Specifications

Antigen ZNF379
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZDHHC9
Gene Accession No. Q9Y397
Gene Alias 6430508G22; 9530098M12Rik; antigen MMSA-1; Asp-His-His-Cys domain containing protein 9; CGI89; CGI-89; CXorf11; DHHC9; DHHC-9; membrane-associated DHHC9 zinc finger protein; MMSA1; MRXSZ; Palmitoyltransferase ZDHHC9; UNQ261/PRO298; ZDHHC10; Zdhhc9; Zinc finger DHHC domain-containing protein 9; zinc finger DHHC-type containing 9; Zinc finger protein 379; Zinc finger protein 380; zinc finger, DHHC domain containing 10; zinc finger, DHHC domain containing 9; zinc finger, DHHC-type containing 9; ZNF379; ZNF380
Gene Symbols ZDHHC9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZNF379/ZDHHC9.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51114
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.