Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF37A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598260
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598260 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598260 Supplier Invitrogen™ Supplier No. PA598260
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF37A Polyclonal Antibody for Western Blot, ELISA

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF37A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZNF37A
Gene Accession No. P17032
Gene Alias KOX21; Zinc finger protein 37A; zinc finger protein 37a (KOX 21); zinc finger protein KOX21; ZNF37; ZNF37A
Gene Symbols ZNF37A
Host Species Rabbit
Immunogen Recombinant Human Zinc finger protein 37A protein (263-386AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7587
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.